Assay Details
| Gene Symbol: hvcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011262.1 | NP_001011262.1 | 4 - 5 | 609 | 64 | |
| XM_012961069.2 | XP_012816523.1 | 4 - 5 | 712 | 64 | ||
| XM_012961072.2 | XP_012816526.1 | 4 - 5 | 665 | 64 | ||
| GenBank mRNA | BC087968.1 | - | 4 - 5 | 609 | 64 | |
More Information
Set Membership: |
|

hvcn1
Genomic Map