Assay Details
| Gene Symbol: vnn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011263.1 | NP_001011263.1 | 5 - 6 | 1267 | 94 | |
| GenBank mRNA | BC087969.1 | - | 5 - 6 | 1267 | 94 | |
More Information
Set Membership: |
|

vnn2
Genomic Map