Assay Details
| Gene Symbol: ahsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011278.1 | NP_001011278.1 | 2 - 3 | 336 | 76 | |
| GenBank mRNA | BC087992.1 | - | 2 - 3 | 336 | 76 | |
More Information
Set Membership: |
|

ahsg
Genomic Map