Assay Details
| Gene Symbol: krt19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203587.1 | NP_988918.1 | 5 - 6 | 1075 | 71 | |
| XM_012971580.2 | XP_012827034.1 | 5 - 6 | 1104 | 71 | ||
| GenBank mRNA | BC061624.1 | - | 5 - 6 | 1075 | 71 | |
| CR761861.2 | - | 5 - 6 | 1039 | 71 | ||
More Information
Set Membership: |
|

krt19
Genomic Map