Assay Details
| Gene Symbol: impact | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011308.1 | NP_001011308.1 | 2 - 3 | 290 | 70 | |
| GenBank mRNA | BC088032.1 | - | 2 - 3 | 290 | 70 | |
| CR760285.2 | - | 2 - 3 | 297 | 70 | ||
More Information
Set Membership: |
|

impact
Genomic Map