Assay Details
| Gene Symbol: atraid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011316.1 | NP_001011316.1 | 469 | 60 | ||
| GenBank mRNA | BC088051.1 | - | 469 | 60 | ||
| CU075626.1 | - | 3 - 4 | 465 | 60 | ||
More Information
Set Membership: |
|

atraid
Genomic Map