Assay Details
| Gene Symbol: mettl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011322.1 | NP_001011322.1 | 7 - 8 | 790 | 85 | |
| GenBank mRNA | BC088068.1 | - | 7 - 8 | 790 | 85 | |
| CU075661.1 | - | 7 - 8 | 988 | 85 | ||
More Information
Set Membership: |
|

mettl2b
Genomic Map