Assay Details
| Gene Symbol: pdcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011382.1 | NP_001011382.1 | 4 - 5 | 305 | 67 | |
| XM_012956868.1 | XP_012812322.1 | 4 - 5 | 377 | 67 | ||
| GenBank mRNA | BC088564.1 | - | 4 - 5 | 305 | 67 | |
| CR761441.2 | - | 4 - 5 | 305 | 67 | ||
More Information
Set Membership: |
|

pdcl3
Genomic Map