Assay Details
| Gene Symbol: adh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011391.1 | NP_001011391.1 | 2 - 3 | 191 | 101 | |
| GenBank mRNA | BC088582.1 | - | 2 - 3 | 191 | 101 | |
More Information
Set Membership: |
|

adh1
Genomic Map