Assay Details
| Gene Symbol: timm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011397.1 | NP_001011397.1 | 2 - 3 | 458 | 88 | |
| GenBank mRNA | BC088600.1 | - | 2 - 3 | 458 | 88 | |
More Information
Set Membership: |
|

timm22
Genomic Map