Assay Details
| Gene Symbol: gyg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011403.1 | NP_001011403.1 | 9 - 10 | 1188 | 80 | |
| XM_012956872.2 | XP_012812326.1 | 6 - 7 | 812 | 80 | ||
| GenBank mRNA | BC088760.1 | - | 9 - 10 | 1188 | 80 | |
More Information
Set Membership: |
|

gyg2
Genomic Map