Assay Details
| Gene Symbol: ndufa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011432.1 | NP_001011432.1 | 6 - 7 | 672 | 65 | |
| GenBank mRNA | BC088072.1 | - | 6 - 7 | 672 | 65 | |
More Information
Set Membership: |
|

ndufa9
Genomic Map