Assay Details
| Gene Symbol: tk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011452.1 | NP_001011452.1 | 6 - 7 | 532 | 78 | |
| GenBank mRNA | BC088552.1 | - | 6 - 7 | 532 | 78 | |
| CR855612.2 | - | 6 - 7 | 525 | 78 | ||
More Information
Set Membership: |
|

tk1
Genomic Map