Assay Details
| Gene Symbol: bud23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011475.1 | NP_001011475.1 | 3 - 4 | 186 | 98 | |
| GenBank mRNA | BC088766.1 | - | 3 - 4 | 186 | 98 | |
More Information
Set Membership: |
|

bud23
Genomic Map