Assay Details
| Gene Symbol: rpl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011478.1 | NP_001011478.1 | 2 - 3 | 332 | 61 | |
| XM_012964627.1 | XP_012820081.1 | 3 - 4 | 371 | 61 | ||
| XM_018094638.1 | XP_017950127.1 | 3 - 4 | 346 | 61 | ||
| XM_018094639.1 | XP_017950128.1 | 3 - 4 | 346 | 61 | ||
| XM_018094640.1 | XP_017950129.1 | 3 - 4 | 346 | 61 | ||
| GenBank mRNA | BC080484.1 | - | 3 - 4 | 345 | 61 | |
| BC088771.1 | - | 2 - 3 | 332 | 61 | ||
| BC166327.2 | - | 2 - 3 | 335 | 61 | ||
| CR761463.2 | - | 2 - 3 | 321 | 61 | ||
More Information
Set Membership: |
|

rpl15
Genomic Map