Assay Details
| Gene Symbol: hck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015722.1 | NP_001015722.1 | 678 | 61 | ||
| GenBank mRNA | BC089654.1 | - | 678 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hck| Gene Symbol: hck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015722.1 | NP_001015722.1 | 678 | 61 | ||
| GenBank mRNA | BC089654.1 | - | 678 | 61 | ||
Set Membership: |
|