Assay Details
| Gene Symbol: tdo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015733.1 | NP_001015733.1 | 4 - 5 | 329 | 84 | |
| GenBank mRNA | BC089667.1 | - | 4 - 5 | 329 | 84 | |
More Information
Set Membership: |
|

tdo2
Genomic Map