Assay Details
| Gene Symbol: zeb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015808.1 | NP_001015808.1 | 6 - 7 | 2522 | 65 | |
| XM_012964650.2 | XP_012820104.1 | 7 - 8 | 2619 | 65 | ||
| XM_012964652.2 | XP_012820106.1 | 7 - 8 | 2620 | 65 | ||
| XM_012964653.2 | XP_012820107.1 | 7 - 8 | 2891 | 65 | ||
| GenBank mRNA | BC090088.1 | - | 6 - 7 | 2522 | 65 | |
More Information
Set Membership: |
|

zeb1
Genomic Map