Assay Details
| Gene Symbol: lsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015935.2 | NP_001015935.1 | 1 - 2 | 118 | 67 | |
| XM_018095387.1 | XP_017950876.1 | 1 - 2 | 342 | 67 | ||
| GenBank mRNA | BC161040.1 | - | 2 - 3 | 172 | 67 | |
| CR926369.2 | - | 1 - 2 | 118 | 67 | ||
More Information
Set Membership: |
|

lsr
Genomic Map