Assay Details
| Gene Symbol: prim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016008.3 | NP_001016008.1 | 3 - 4 | 397 | 78 | |
| GenBank mRNA | BC170847.1 | - | 3 - 4 | 387 | 78 | |
| BC170875.1 | - | 3 - 4 | 387 | 78 | ||
| CR848228.2 | - | 3 - 4 | 399 | 78 | ||
More Information
Set Membership: |
|

prim2
Genomic Map