Assay Details
| Gene Symbol: rgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016013.3 | NP_001016013.1 | 3 - 4 | 363 | 77 | |
| GenBank mRNA | BC135113.1 | - | 3 - 4 | 380 | 77 | |
| CR848213.2 | - | 3 - 4 | 366 | 77 | ||
More Information
Set Membership: |
|

rgr
Genomic Map