Assay Details
| Gene Symbol: ube2l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016063.2 | NP_001016063.1 | 3 - 4 | 328 | 60 | |
| GenBank mRNA | BC091621.1 | - | 3 - 4 | 328 | 60 | |
| CR762273.2 | - | 3 - 4 | 328 | 60 | ||
More Information
Set Membership: |
|

ube2l3
Genomic Map