Assay Details
| Gene Symbol: cyp51a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016194.2 | NP_001016194.1 | 10 - 11 | 1728 | 68 | |
| GenBank mRNA | BC135319.1 | - | 10 - 11 | 1582 | 68 | |
| CR926353.2 | - | 10 - 11 | 1728 | 68 | ||
More Information
Set Membership: |
|

cyp51a1
Genomic Map