Assay Details
| Gene Symbol: jmjd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016203.2 | NP_001016203.1 | 1 - 2 | 132 | 99 | |
| GenBank mRNA | CR926342.2 | - | 1 - 2 | 132 | 99 | |
More Information
Set Membership: |
|

jmjd6
Genomic Map