Assay Details
| Gene Symbol: coq4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016225.1 | NP_001016225.1 | 2 - 3 | 293 | 72 | |
| GenBank mRNA | BC161730.1 | - | 2 - 3 | 329 | 72 | |
| BC171035.1 | - | 2 - 3 | 288 | 72 | ||
| CR926300.1 | - | 2 - 3 | 293 | 72 | ||
More Information
Set Membership: |
|

coq4
Genomic Map