Assay Details
| Gene Symbol: gtf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016277.2 | NP_001016277.1 | 3 - 4 | 286 | 96 | |
| GenBank mRNA | BC160469.1 | - | 3 - 4 | 279 | 96 | |
| CR761615.2 | - | 3 - 4 | 286 | 96 | ||
More Information
Set Membership: |
|

gtf2b
Genomic Map