Assay Details
| Gene Symbol: ogfod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016322.2 | NP_001016322.1 | 3 - 4 | 394 | 65 | |
| GenBank mRNA | BC118748.1 | - | 3 - 4 | 366 | 65 | |
| CR761454.2 | - | 3 - 4 | 394 | 65 | ||
More Information
Set Membership: |
|

ogfod1
Genomic Map