Assay Details
| Gene Symbol: actr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016472.1 | NP_001016472.1 | 4 - 5 | 418 | 63 | |
| XM_012959024.2 | XP_012814478.1 | 4 - 5 | 587 | 63 | ||
| GenBank mRNA | BC121585.1 | - | 4 - 5 | 362 | 63 | |
| BC158147.1 | - | 4 - 5 | 566 | 63 | ||
| BC170604.1 | - | 4 - 5 | 418 | 63 | ||
| BC170981.1 | - | 4 - 5 | 418 | 63 | ||
| CR760917.1 | - | 4 - 5 | 418 | 63 | ||
More Information
Set Membership: |
|

actr6
Genomic Map