Assay Details
| Gene Symbol: tfb1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016494.2 | NP_001016494.1 | 4 - 5 | 439 | 87 | |
| XM_012971770.2 | XP_012827224.1 | 4 - 5 | 714 | 87 | ||
| GenBank mRNA | BC160475.1 | - | 4 - 5 | 465 | 87 | |
| BC170821.1 | - | 4 - 5 | 433 | 87 | ||
| BC170849.1 | - | 4 - 5 | 433 | 87 | ||
| CR760823.2 | - | 4 - 5 | 439 | 87 | ||
More Information
Set Membership: |
|

tfb1m
Genomic Map