Assay Details
| Gene Symbol: mespb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016653.1 | NP_001016653.1 | 1 - 2 | 819 | 84 | |
| GenBank mRNA | CR760340.1 | - | 1 - 2 | 819 | 84 | |
More Information
Set Membership: |
|

mespb
Genomic Map