Assay Details
| Gene Symbol: tnfaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016809.2 | NP_001016809.1 | 3 - 4 | 545 | 74 | |
| XM_012957000.2 | XP_012812454.1 | 3 - 4 | 644 | 74 | ||
| XM_012957001.2 | XP_012812455.1 | 3 - 4 | 532 | 74 | ||
| XM_018091268.1 | XP_017946757.1 | 4 - 5 | 820 | 74 | ||
| GenBank mRNA | BC135236.1 | - | 2 - 3 | 507 | 74 | |
| CR855581.2 | - | 3 - 4 | 545 | 74 | ||
More Information
Set Membership: |
|

tnfaip1
Genomic Map