Assay Details
| Gene Symbol: gdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016819.2 | NP_001016819.1 | 1 - 2 | 275 | 67 | |
| GenBank mRNA | BC161554.1 | - | 1 - 2 | 283 | 67 | |
| CR855548.2 | - | 1 - 2 | 275 | 67 | ||
More Information
Set Membership: |
|

gdf1
Genomic Map