Assay Details
Gene Symbol: bcs1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016865.3 | NP_001016865.1 | 7 - 8 | 1140 | 100 | |
XM_012970440.2 | XP_012825894.1 | 7 - 8 | 1143 | 100 | ||
GenBank mRNA | CR848645.2 | - | 7 - 8 | 1142 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
