Assay Details
| Gene Symbol: prelid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016922.2 | NP_001016922.1 | 5 - 6 | 506 | 72 | |
| GenBank mRNA | BC091072.1 | - | 5 - 6 | 506 | 72 | |
| CR848108.2 | - | 5 - 6 | 538 | 72 | ||
More Information
Set Membership: |
|

prelid2
Genomic Map