Assay Details
| Gene Symbol: vps33a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016935.2 | NP_001016935.1 | 1 - 2 | 100 | 73 | |
| XM_012963077.2 | XP_012818531.1 | 1 - 2 | 248 | 73 | ||
| XM_018093616.1 | XP_017949105.1 | 1 - 2 | 248 | 73 | ||
| GenBank mRNA | BC127292.1 | - | 1 - 2 | 102 | 73 | |
| CR762099.2 | - | 1 - 2 | 100 | 73 | ||
More Information
Set Membership: |
|

vps33a
Genomic Map