Assay Details
| Gene Symbol: pak1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017054.3 | NP_001017054.2 | 1 - 2 | 132 | 93 | |
| GenBank mRNA | BC135557.1 | - | 1 - 2 | 99 | 93 | |
| CR761765.2 | - | 1 - 2 | 127 | 93 | ||
| CX331845.2 | - | 1 - 2 | 132 | 93 | ||
More Information
Set Membership: |
|

pak1ip1
Genomic Map