Assay Details
| Gene Symbol: slc25a21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017069.2 | NP_001017069.1 | 8 - 9 | 691 | 57 | |
| XM_012968463.2 | XP_012823917.1 | 8 - 9 | 690 | 57 | ||
| XM_018096274.1 | XP_017951763.1 | 7 - 8 | 679 | 57 | ||
| GenBank mRNA | CR761553.2 | - | 8 - 9 | 691 | 57 | |
More Information
Set Membership: |
|

slc25a21
Genomic Map