Assay Details
| Gene Symbol: srd5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017113.2 | NP_001017113.1 | 4 - 5 | 763 | 70 | |
| XM_018093906.1 | XP_017949395.1 | 4 - 5 | 1010 | 70 | ||
| GenBank mRNA | CR761203.2 | - | 4 - 5 | 763 | 70 | |
More Information
Set Membership: |
|

srd5a2
Genomic Map