Assay Details
| Gene Symbol: ptpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017121.3 | NP_001017121.2 | 2 - 3 | 254 | 64 | |
| GenBank mRNA | BC161039.1 | - | 2 - 3 | 254 | 64 | |
| CR761155.2 | - | 2 - 3 | 222 | 64 | ||
More Information
Set Membership: |
|

ptpn1
Genomic Map