Assay Details
| Gene Symbol: dap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017164.2 | NP_001017164.1 | 1 - 2 | 95 | 62 | |
| GenBank mRNA | BC096501.1 | - | 1 - 2 | 95 | 62 | |
| CR760800.2 | - | 1 - 2 | 115 | 62 | ||
More Information
Set Membership: |
|

dap
Genomic Map