Assay Details
| Gene Symbol: rab20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017295.2 | NP_001017295.1 | 1 - 2 | 254 | 63 | |
| XM_012957051.2 | XP_012812505.1 | 2 - 3 | 335 | 63 | ||
| XM_012957053.2 | XP_012812507.1 | 2 - 3 | 290 | 63 | ||
| XM_012957054.1 | XP_012812508.1 | 2 - 3 | 262 | 63 | ||
| XM_012957055.2 | XP_012812509.1 | 2 - 3 | 261 | 63 | ||
| GenBank mRNA | BC121519.1 | - | 2 - 3 | 261 | 63 | |
| CR855629.2 | - | 1 - 2 | 254 | 63 | ||
More Information
Set Membership: |
|

rab20
Genomic Map