Assay Details
| Gene Symbol: mmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030330.1 | NP_001025501.1 | 9 - 10 | 1330 | 75 | |
| GenBank mRNA | BC080175.1 | - | 9 - 10 | 1336 | 75 | |
| BC161475.1 | - | 9 - 10 | 1323 | 75 | ||
| CR760257.1 | - | 9 - 10 | 1330 | 75 | ||
More Information
Set Membership: |
|

mmp1
Genomic Map