Assay Details
| Gene Symbol: galnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030376.1 | NP_001025547.1 | 5 - 6 | 896 | 72 | |
| GenBank mRNA | BC090583.1 | - | 5 - 6 | 896 | 72 | |
More Information
Set Membership: |
|

galnt1
Genomic Map