Assay Details
| Gene Symbol: hibadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030433.1 | NP_001025604.1 | 2 - 3 | 246 | 60 | |
| GenBank mRNA | BC091056.1 | - | 2 - 3 | 246 | 60 | |
More Information
Set Membership: |
|

hibadh
Genomic Map