Assay Details
| Gene Symbol: rpl3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030441.1 | NP_001025612.1 | 2 - 3 | 239 | 59 | |
| GenBank mRNA | BC091070.1 | - | 2 - 3 | 239 | 59 | |
More Information
Set Membership: |
|

rpl3l
Genomic Map