Assay Details
| Gene Symbol: matn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030442.1 | NP_001025613.1 | 2 - 3 | 447 | 61 | |
| XM_018091297.1 | XP_017946786.1 | 2 - 3 | 480 | 61 | ||
| GenBank mRNA | BC091071.1 | - | 2 - 3 | 447 | 61 | |
More Information
Set Membership: |
|

matn1
Genomic Map