Assay Details
| Gene Symbol: ezr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030458.1 | NP_001025629.1 | 13 - 14 | 1735 | 64 | |
| GenBank mRNA | BC091578.1 | - | 13 - 14 | 1735 | 64 | |
More Information
Set Membership: |
|

ezr
Genomic Map