Assay Details
| Gene Symbol: p3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030459.1 | NP_001025630.1 | 1 - 2 | 1020 | 73 | |
| GenBank mRNA | BC091581.1 | - | 1 - 2 | 1020 | 73 | |
More Information
Set Membership: |
|

p3h4
Genomic Map