Assay Details
| Gene Symbol: c9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030461.2 | NP_001025632.2 | 3 - 4 | 689 | 82 | |
| GenBank mRNA | BC091587.1 | - | 3 - 4 | 661 | 82 | |
More Information
Set Membership: |
|

c9orf64
Genomic Map