Assay Details
| Gene Symbol: c9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030461.2 | NP_001025632.2 | 4 - 5 | 889 | 101 | |
| GenBank mRNA | BC091587.1 | - | 4 - 5 | 861 | 101 | |
| DN096759.1 | - | 1 - 2 | 94 | 101 | ||
More Information
Set Membership: |
|

c9orf64
Genomic Map